Merck
  • Palladium-catalyzed cross-coupling reactions of silanolates: a paradigm shift in silicon-based cross-coupling reactions.
Palladium-catalyzed cross-coupling reactions of silanolates: a paradigm shift in silicon-based cross-coupling reactions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2006-05-04)
Scott E Denmark, John D Baird
ABSTRACT
This paper chronicles the conceptual development, proof of principle experiments, and recent advances in the palladium-catalyzed cross-coupling reactions of the conjugate bases of organosilanols. The discovery that led to the design and refinement of this process represents a classical illustration of how mechanistic studies can provide a fertile ground for the invention of new reactions. On the basis of a working hypothesis (which ultimately proved to be incorrect) and the desire to effect silicon-based cross-coupling without the agency of fluoride activation, a mild and practical palladium-catalyzed cross-coupling of alkenyl-, aryl-, and heteroaryl silanolates has been developed. The mechanistic underpinnings, methodological extensions, and the successful applications of this technology to the synthesis of complex molecules are described.
Sigma-Aldrich
Potassium tert-butoxide, reagent grade, ≥98%
Sigma-Aldrich
Potassium tert-butoxide solution, 1.0 M in THF
Sigma-Aldrich
Potassium tert-butoxide, sublimed grade, 99.99% trace metals basis