Merck
  • Pharmacokinetics of 6-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol and conjugate metabolites in healthy human subjects.
Pharmacokinetics of 6-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol and conjugate metabolites in healthy human subjects.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2008-08-19)
Suzanna M Zick, Zora Djuric, Mack T Ruffin, Amie J Litzinger, Daniel P Normolle, Sara Alrawi, Meihua Rose Feng, Dean E Brenner
ABSTRACT
Ginger shows promising anticancer properties. No research has examined the pharmacokinetics of the ginger constituents 6-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol in humans. We conducted a clinical trial with 6-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol, examining the pharmacokinetics and tolerability of these analytes and their conjugate metabolites. Human volunteers were given ginger at doses from 100 mg to 2.0 g (N = 27), and blood samples were obtained at 15 minutes to 72 hours after a single p.o. dose. The participants were allocated in a dose-escalation manner starting with 100 mg. There was a total of three participants at each dose except for 1.0 g (N = 6) and 2.0 g (N = 9). No participant had detectable free 6-gingerol, 8-gingerol, 10-gingerol, or 6-shogaol, but 6-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol glucuronides were detected. The 6-gingerol sulfate conjugate was detected above the 1.0-g dose, but there were no detectable 10-gingerol or 6-shogaol sulfates except for one participant with detectable 8-gingerol sulfate. The C(max) and area under the curve values (mean +/- SE) estimated for the 2.0-g dose are 0.85 +/- 0.43, 0.23 +/- 0.16, 0.53 +/- 0.40, and 0.15 +/- 0.12 microg/mL; and 65.6.33 +/- 44.4, 18.1 +/- 20.3, 50.1 +/- 49.3, and 10.9 +/- 13.0 microg x hr/mL for 6-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol. The corresponding t(max) values are 65.6 +/- 44.4, 73.1 +/- 29.4, 75.0 +/- 27.8, and 65.6 +/- 22.6 minutes, and the analytes had elimination half-lives <2 hours. The 8-gingerol, 10-gingerol, and 6-shogaol conjugates were present as either glucuronide or sulfate conjugates, not as mixed conjugates, although 6-gingerol and 10-gingerol were an exception. Six-gingerol, 8-gingerol, 10-gingerol, and 6-shogaol are absorbed after p.o. dosing and can be detected as glucuronide and sulfate conjugates.
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
Supelco
[8]-Gingerol, analytical standard