Merck
  • Members of the E2D (UbcH5) family mediate the ubiquitination of the conserved cysteine of Pex5p, the peroxisomal import receptor.
Members of the E2D (UbcH5) family mediate the ubiquitination of the conserved cysteine of Pex5p, the peroxisomal import receptor.
The Journal of biological chemistry (2008-03-25)
Cláudia P Grou, Andreia F Carvalho, Manuel P Pinto, Sebastian Wiese, Heike Piechura, Helmut E Meyer, Bettina Warscheid, Clara Sá-Miranda, Jorge E Azevedo
ABSTRACT
According to current models of peroxisomal biogenesis, newly synthesized peroxisomal matrix proteins are transported into the organelle by Pex5p. Pex5p recognizes these proteins in the cytosol, mediates their membrane translocation, and is exported back into the cytosol in an ATP-dependent manner. We have previously shown that export of Pex5p is preceded by (and requires) monoubiquitination of a conserved cysteine residue present at its N terminus. In yeasts, and probably also in plants, ubiquitination of Pex5p is mediated by a specialized ubiquitin-conjugating enzyme, Pex4p. In mammals, the identity of this enzyme has remained unknown for many years. Here, we provide evidence suggesting that E2D1/2/3 (UbcH5a/b/c) are the mammalian functional counterparts of yeast/plant Pex4p. The mechanistic implications of these findings are discussed.
Product Description
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 20 ea (5 bags × 4 plates)
Eppendorf® Protein LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Protein LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® DNA LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® DNA LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Protein LoBind Microplate 384/V-PP, PCR clean, V-bottom clear wells, white border, pack of 240 ea (10 bags × 24 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 120 ea (10 bags × 12 plates)
Eppendorf® DNA LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)