Merck
  • IRF5 promotes glycolysis in the progression of hepatocellular carcinoma and is regulated by TRIM35.
IRF5 promotes glycolysis in the progression of hepatocellular carcinoma and is regulated by TRIM35.
Journal of digestive diseases (2023-08-18)
Ying Fang, Zhi Hui Lu, Bang Zhong Liu, Nan Li, Ming Zhen Yang, Ping Wang
ABSTRACT
The interferon regulatory factor (IRF) family of proteins are involved in tumor progression. However, the role of IRF5 in tumorigenesis remains unknown. In this study we aimed to elucidate the functions of IRF5 in the progression of hepatocellular carcinoma (HCC). IRF5 expression in HCC was analyzed through quantitative polymerase chain reaction (qPCR), western blot, and immunohistochemistry (IHC), etc. The Cell Counting Kit 8 (CCK8) assay, anchorage-independent assay, and EdU assay were used to evaluate the role of IRF5. The molecular mechanisms were studied by analyzing the metabolites with mass spectrum and immunoprecipitation. IRF5 was upregulated in HCC. Interfering with IRF5 inhibited the proliferation and tumorigenic potential of HCC cells. When studying the molecular mechanism, IRF5 was found to upregulate the expression of lactate dehydrogenase A (LDHA) and promoted glycolysis. Additionally, tripartite motif containing 35 (TRIM35) interacted with IRF5, promoting its ubiquitination and degradation. In the clinically obtained HCC samples, TRIM35 was negatively correlated with the expression of IRF5. These findings reveal the oncogenic function of IRF5 in the progression of HCC by enhancing glycolysis, further supporting the potential of IRF5 as a viable target for HCC therapy.
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-IRF5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TRIM35 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® BioM2 antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution