Merck
  • Novel CDK12/13 Inhibitors AU-15506 and AU-16770 Are Potent Anti-Cancer Agents in EGFR Mutant Lung Adenocarcinoma with and without Osimertinib Resistance.
Novel CDK12/13 Inhibitors AU-15506 and AU-16770 Are Potent Anti-Cancer Agents in EGFR Mutant Lung Adenocarcinoma with and without Osimertinib Resistance.
Cancers (2023-05-16)
Tapan K Maity, Eun Young Kim, Constance M Cultraro, Abhilash Venugopalan, Leena Khare, Ramulu Poddutoori, Sivapriya Marappan, Samiulla D Syed, William G Telford, Susanta Samajdar, Murali Ramachandra, Udayan Guha
ABSTRACT
Osimertinib is a third-generation epidermal growth factor receptor and tyrosine kinase inhibitor (EGFR-TKI) approved for the treatment of lung adenocarcinoma patients harboring EGFR mutations. However, acquired resistance to this targeted therapy is inevitable, leading to disease relapse within a few years. Therefore, understanding the molecular mechanisms of osimertinib resistance and identifying novel targets to overcome such resistance are unmet needs of cancer patients. Here, we investigated the efficacy of two novel CDK12/13 inhibitors, AU-15506 and AU-16770, in osimertinib-resistant EGFR mutant lung adenocarcinoma cells in culture and xenograft models in vivo. We demonstrate that these drugs, either alone or in combination with osimertinib, are potent inhibitors of osimertinib-resistant as well as -sensitive lung adenocarcinoma cells in culture. Interestingly, only the CDK12/13 inhibitor in combination with osimertinib, although not as monotherapy, suppresses the growth of resistant tumors in xenograft models in vivo. Taken together, the results of this study suggest that inhibition of CDK12/13 in combination with osimertinib has the potential to overcome osimertinib resistance in EGFR mutant lung adenocarcinoma patients.
Sigma-Aldrich
Anti-CDK12 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho RNA Pol II (Ser2), clone 3E7C7 Antibody, clone 3E7C7, from rat
Sigma-Aldrich
Anti-phospho RNA Pol II (Ser5), clone 1H4B6 Antibody, clone 1H4B6, from rat
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial