Merck
  • Adeno-Associated Virus-Mediated Gene Therapy in the Mashlool, Atp1a3Mashl/+, Mouse Model of Alternating Hemiplegia of Childhood.
Adeno-Associated Virus-Mediated Gene Therapy in the Mashlool, Atp1a3Mashl/+, Mouse Model of Alternating Hemiplegia of Childhood.
Human gene therapy (2021-02-13)
Arsen S Hunanyan, Boris Kantor, Ram S Puranam, Courtney Elliott, Angela McCall, Justin Dhindsa, Promila Pagadala, Keri Wallace, Jordan Poe, Talha Gunduz, Aravind Asokan, Dwight D Koeberl, Mai K ElMallah, Mohamad A Mikati
ABSTRACT
Alternating Hemiplegia of Childhood (AHC) is a devastating autosomal dominant disorder caused by ATP1A3 mutations, resulting in severe hemiplegia and dystonia spells, ataxia, debilitating disabilities, and premature death. Here, we determine the effects of delivering an extra copy of the normal gene in a mouse model carrying the most common mutation causing AHC in humans, the D801N mutation. We used an adeno-associated virus serotype 9 (AAV9) vector expressing the human ATP1A3 gene under the control of a human Synapsin promoter. We first demonstrated that intracerebroventricular (ICV) injection of this vector in wild-type mice on postnatal day 10 (P10) results in increases in ouabain-sensitive ATPase activity and in expression of reporter genes in targeted brain regions. We then tested this vector in mutant mice. Simultaneous intracisterna magna and bilateral ICV injections of this vector at P10 resulted, at P40, in reduction of inducible hemiplegia spells, improvement in balance beam test performance, and prolonged survival of treated mutant mice up to P70. Our study demonstrates, as a proof of concept, that gene therapy can induce favorable effects in a disease caused by a mutation of the gene of a protein that is, at the same time, an ATPase enzyme, a pump, and a signal transduction factor.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ATPase/GTPase Activity Assay Kit, sufficient for 200 colorimetric tests
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 9E10, Alexa Fluor 488 conjugate, clone 9E10, Upstate®, from mouse