Merck
  • Gene regulatory networks controlling temporal patterning, neurogenesis, and cell-fate specification in mammalian retina.
Gene regulatory networks controlling temporal patterning, neurogenesis, and cell-fate specification in mammalian retina.
Cell reports (2021-11-18)
Pin Lyu, Thanh Hoang, Clayton P Santiago, Eric D Thomas, Andrew E Timms, Haley Appel, Megan Gimmen, Nguyet Le, Lizhi Jiang, Dong Won Kim, Siqi Chen, David F Espinoza, Ariel E Telger, Kurt Weir, Brian S Clark, Timothy J Cherry, Jiang Qian, Seth Blackshaw
ABSTRACT
Gene regulatory networks (GRNs), consisting of transcription factors and their target sites, control neurogenesis and cell-fate specification in the developing central nervous system. In this study, we use integrated single-cell RNA and single-cell ATAC sequencing (scATAC-seq) analysis in developing mouse and human retina to identify multiple interconnected, evolutionarily conserved GRNs composed of cell-type-specific transcription factors that both activate genes within their own network and inhibit genes in other networks. These GRNs control temporal patterning in primary progenitors, regulate transition from primary to neurogenic progenitors, and drive specification of each major retinal cell type. We confirm that NFI transcription factors selectively activate expression of genes promoting late-stage temporal identity in primary retinal progenitors and identify other transcription factors that regulate rod photoreceptor specification in postnatal retina. This study inventories cis- and trans-acting factors that control retinal development and can guide cell-based therapies aimed at replacing retinal neurons lost to disease.
Sigma-Aldrich
Anti-NFIA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Monoclonal Anti-NFIX antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution