Merck
  • Galectin-9 regulates the threshold of B cell activation and autoimmunity.
Galectin-9 regulates the threshold of B cell activation and autoimmunity.
eLife (2021-08-10)
Logan K Smith, Kareem Fawaz, Bebhinn Treanor
ABSTRACT
Despite the mechanisms of central and peripheral tolerance, the mature B cell compartment contains cells reactive for self-antigen. How these cells are poised not to respond and the mechanisms that restrain B cell responses to low-affinity endogenous antigens are not fully understood. Here, we demonstrate a critical role for the glycan-binding protein galectin-9 in setting the threshold of B cell activation and that loss of this regulatory network is sufficient to drive spontaneous autoimmunity. We further demonstrate a critical role for galectin-9 in restraining not only conventional B-2 B cells, but also innate-like B-1a cells. We show that galectin-9-deficient mice have an expanded population of B-1a cells and increased titers of B-1a-derived autoantibodies. Mechanistically, we demonstrate that galectin-9 regulates BCR and distinct TLR responses in B-1a cells, but not B-1b cells, by regulating the interaction between BCR and TLRs with the regulatory molecules CD5 and CD180, respectively. In the absence of galectin-9, B-1a cells are more readily activated and secrete increased titers of autoantibodies that facilitate autoantigen delivery to the spleen, driving autoimmune responses.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Phosphocholine chloride calcium salt tetrahydrate, Sigma Grade
Roche
Lysozyme, from hen egg white
Sigma-Aldrich
β-Lactose, ≤30% α-anomer basis, ≥99% total lactose basis