Merck
  • Use of a 3-D dispersion model for calculation of distribution of horse allergen and odor around horse facilities.
Use of a 3-D dispersion model for calculation of distribution of horse allergen and odor around horse facilities.
International journal of environmental research and public health (2014-04-03)
Marie Haeger-Eugensson, Martin Ferm, Lena Elfman
ABSTRACT
The interest in equestrian sports has increased substantially during the last decades, resulting in increased number of horse facilities around urban areas. In Sweden, new guidelines for safe distance have been decided based on the size of the horse facility (e.g., number of horses) and local conditions, such as topography and meteorology. There is therefore an increasing need to estimate dispersion of horse allergens to be used, for example, in the planning processes for new residential areas in the vicinity of horse facilities. The aim of this study was to develop a method for calculating short- and long-term emissions and dispersion of horse allergen and odor around horse facilities. First, a method was developed to estimate horse allergen and odor emissions at hourly resolution based on field measurements. Secondly, these emission factors were used to calculate concentrations of horse allergen and odor by using 3-D dispersion modeling. Results from these calculations showed that horse allergens spread up to about 200 m, after which concentration levels were very low (<2 U/m³). Approximately 10% of a study-group detected the smell of manure at 60m, while the majority--80%-90%--detected smell at 60 m or shorter distance from the manure heap. Modeling enabled horse allergen exposure concentrations to be determined with good time resolution.
Product Description
Millipore
PTFE Membrane Filter, 5.0 μm Pore Size, Mitex, filter diam. 47 mm, hydrophobic, white