Merck
  • A small molecule compound with an indole moiety inhibits the main protease of SARS-CoV-2 and blocks virus replication.
A small molecule compound with an indole moiety inhibits the main protease of SARS-CoV-2 and blocks virus replication.
Nature communications (2021-01-30)
Shin-Ichiro Hattori, Nobuyo Higashi-Kuwata, Hironori Hayashi, Srinivasa Rao Allu, Jakka Raghavaiah, Haydar Bulut, Debananda Das, Brandon J Anson, Emma K Lendy, Yuki Takamatsu, Nobutoki Takamune, Naoki Kishimoto, Kazutaka Murayama, Kazuya Hasegawa, Mi Li, David A Davis, Eiichi N Kodama, Robert Yarchoan, Alexander Wlodawer, Shogo Misumi, Andrew D Mesecar, Arun K Ghosh, Hiroaki Mitsuya
ABSTRACT
Except remdesivir, no specific antivirals for SARS-CoV-2 infection are currently available. Here, we characterize two small-molecule-compounds, named GRL-1720 and 5h, containing an indoline and indole moiety, respectively, which target the SARS-CoV-2 main protease (Mpro). We use VeroE6 cell-based assays with RNA-qPCR, cytopathic assays, and immunocytochemistry and show both compounds to block the infectivity of SARS-CoV-2 with EC50 values of 15 ± 4 and 4.2 ± 0.7 μM for GRL-1720 and 5h, respectively. Remdesivir permitted viral breakthrough at high concentrations; however, compound 5h completely blocks SARS-CoV-2 infection in vitro without viral breakthrough or detectable cytotoxicity. Combination of 5h and remdesivir exhibits synergism against SARS-CoV-2. Additional X-ray structural analysis show that 5h forms a covalent bond with Mpro and makes polar interactions with multiple active site amino acid residues. The present data suggest that 5h might serve as a lead Mpro inhibitor for the development of therapeutics for SARS-CoV-2 infection.
Sigma-Aldrich
GRL-1720, ≥98% (HPLC)
Sigma-Aldrich
Mpro inhibitor 5h, ≥98% (HPLC)
Sigma-Aldrich
Atazanavir, ≥98% (HPLC)