Merck
  • Targeting folate receptor β positive tumor-associated macrophages in lung cancer with a folate-modified liposomal complex.
Targeting folate receptor β positive tumor-associated macrophages in lung cancer with a folate-modified liposomal complex.
Signal transduction and targeted therapy (2020-04-17)
Yan Tie, Heng Zheng, Zhiyao He, Jingyun Yang, Bin Shao, Li Liu, Min Luo, Xia Yuan, Yu Liu, Xiangxian Zhang, Hongyi Li, Min Wu, Xiawei Wei
ABSTRACT
Tumor-associated macrophages (TAMs) facilitate cancer progression by promoting tumor invasion, angiogenesis, metastasis, inflammatory responses, and immunosuppression. Folate receptor β (FRβ) is overexpressed in TAMs. However, the clinical significance of FRβ-positive macrophages in lung cancer remains poorly understood. In this study, we verified that FRβ overexpression in lung cancer TAMs was associated with poor prognosis. We utilized a folate-modified lipoplex comprising a folate-modified liposome (F-PLP) delivering a BIM-S plasmid to target both lung cancer cells and FRβ-positive macrophages in the tumor microenvironment. Transfection of LL/2 cells and MH-S cells with F-PLP/pBIM induced cell apoptosis. Injection of F-PLP/pBIM into LL/2 and A549 lung cancer models significantly depleted FRβ-positive macrophages and reduced tumor growth. Treatment of tumor-bearing mice with F-PLP/pBIM significantly inhibited tumor growth in vivo by inducing tumor cell and macrophage apoptosis, reducing tumor proliferation, and inhibiting tumor angiogenesis. In addition, a preliminary safety evaluation demonstrated a good safety profile of F-PLP/pBIM as a gene therapy administered intravenously. This work describes a novel application of lipoplexes in lung cancer targeted therapy that influences the tumor microenvironment by targeting TAMs.
Sigma-Aldrich
NanoFabTx - Natural Folate Lipid Mix, for synthesis of folate functionalized liposomes for targeted delivery
Sigma-Aldrich
ANTI-FOLR2(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
NanoFabTx - Synthetic Folate Lipid Mix, for synthesis of folate functionalized liposomes for targeted delivery