Merck
  • The Potential of Adjusting Water Bolus Liquid Properties for Economic and Precise MR Thermometry Guided Radiofrequency Hyperthermia.
The Potential of Adjusting Water Bolus Liquid Properties for Economic and Precise MR Thermometry Guided Radiofrequency Hyperthermia.
Sensors (Basel, Switzerland) (2020-05-28)
Kemal Sumser, Gennaro G Bellizzi, Gerard C van Rhoon, Margarethus M Paulides
ABSTRACT
The potential of MR thermometry (MRT) fostered the development of MRI compatible radiofrequency (RF) hyperthermia devices. Such device integration creates major technological challenges and a crucial point for image quality is the water bolus (WB). The WB is located between the patient body and external sources to both couple electromagnetic energy and to cool the patient skin. However, the WB causes MRT errors and unnecessarily large field of view. In this work, we studied making the WB MRI transparent by an optimal concentration of compounds capable of modifying T 2 * relaxation without an impact on the efficiency of RF heating. Three different T 2 * reducing compounds were investigated, namely CuSO 4 , MnCl 2 , and Fe 3 O 4 . First, electromagnetic properties and T 2 * relaxation rates at 1.5 T were measured. Next, through multi-physics simulations, the predicted effect on the RF-power deposition pattern was evaluated and MRT precision was experimentally assessed. Our results identified 5 mM Fe 3 O 4 solution as optimal since it does not alter the RF-power level needed and improved MRT precision from 0.39 ∘ C to 0.09 ∘ C. MnCl 2 showed a similar MRT improvement, but caused unacceptable RF-power losses. We conclude that adding Fe 3 O 4 has significant potential to improve RF hyperthermia treatment monitoring under MR guidance.
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Pricing and availability is not currently available.
Supelco
Copper(II) sulfate anhydrous, for analysis EMSURE®
Pricing and availability is not currently available.