Merck
  • Arachidonic Acid Promotes Intestinal Regeneration by Activating WNT Signaling.
Arachidonic Acid Promotes Intestinal Regeneration by Activating WNT Signaling.
Stem cell reports (2020-07-11)
Qingyu Wang, Yingying Lin, Xiaole Sheng, Jiuzhi Xu, Xuening Hou, Yuan Li, Hao Zhang, Huiyuan Guo, Zhengquan Yu, Fazheng Ren
ABSTRACT
Intestinal regeneration is crucial for functional restoration after injury, and nutritional molecules can play an important role in this process. Here, we found that arachidonic acid (AA) serves as a direct proliferation promoter of intestinal epithelial cells that facilitates small intestinal regeneration in both three-dimensional cultured organoids and mouse models. As shown in the study, during post-irradiation regeneration, AA positively regulates intestinal epithelial cell proliferation by upregulating the expression of Ascl2 and activating WNT signaling, but negatively regulates intestinal epithelial cell differentiation. AA acts as a delicate regulator that efficiently facilitates epithelial tissue repair by activating radiation-resistant Msi1+ cells rather than Lgr5+ cells, which are extensively considered WNT-activated crypt base stem cells. Additionally, short-term AA treatment maintains optimal intestinal epithelial homeostasis under physiological conditions. As a result, AA treatment can be considered a potential therapy for irradiation injury repair and tissue regeneration.
Sigma-Aldrich
Anti-Achaete Scute homolog 2 Antibody, clone 8F1, clone 8F1, from mouse