Merck
  • Cloning and characterization of two Lactobacillus casei genes encoding a cystathionine lyase.
Cloning and characterization of two Lactobacillus casei genes encoding a cystathionine lyase.
Applied and environmental microbiology (2007-11-13)
Stefan Irmler, Sylvie Raboud, Beata Beisert, Doris Rauhut, Hélène Berthoud
ABSTRACT
Volatile sulfur compounds are key flavor compounds in several cheese types. To better understand the metabolism of sulfur-containing amino acids, which certainly plays a key role in the release of volatile sulfur compounds, we searched the genome database of Lactobacillus casei ATCC 334 for genes encoding putative homologs of enzymes known to degrade cysteine, cystathionine, and methionine. The search revealed that L. casei possesses two genes that putatively encode a cystathionine beta-lyase (CBL; EC 4.4.1.8). The enzyme has been implicated in the degradation of not only cystathionine but also cysteine and methionine. Recombinant CBL proteins catalyzed the degradation of L-cystathionine, O-succinyl-L-homoserine, L-cysteine, L-serine, and L-methionine to form alpha-keto acid, hydrogen sulfide, or methanethiol. The two enzymes showed notable differences in substrate specificity and pH optimum.
Sigma-Aldrich
O-Succinyl-L-homoserine