Merck
  • Targeting RNA structure in SMN2 reverses spinal muscular atrophy molecular phenotypes.
Targeting RNA structure in SMN2 reverses spinal muscular atrophy molecular phenotypes.
Nature communications (2018-05-26)
Amparo Garcia-Lopez, Francesca Tessaro, Hendrik R A Jonker, Anna Wacker, Christian Richter, Arnaud Comte, Nikolaos Berntenis, Roland Schmucki, Klas Hatje, Olivier Petermann, Gianpaolo Chiriano, Remo Perozzo, Daniel Sciarra, Piotr Konieczny, Ignacio Faustino, Guy Fournet, Modesto Orozco, Ruben Artero, Friedrich Metzger, Martin Ebeling, Peter Goekjian, Benoît Joseph, Harald Schwalbe, Leonardo Scapozza
ABSTRACT
Modification of SMN2 exon 7 (E7) splicing is a validated therapeutic strategy against spinal muscular atrophy (SMA). However, a target-based approach to identify small-molecule E7 splicing modifiers has not been attempted, which could reveal novel therapies with improved mechanistic insight. Here, we chose as a target the stem-loop RNA structure TSL2, which overlaps with the 5' splicing site of E7. A small-molecule TSL2-binding compound, homocarbonyltopsentin (PK4C9), was identified that increases E7 splicing to therapeutic levels and rescues downstream molecular alterations in SMA cells. High-resolution NMR combined with molecular modelling revealed that PK4C9 binds to pentaloop conformations of TSL2 and promotes a shift to triloop conformations that display enhanced E7 splicing. Collectively, our study validates TSL2 as a target for small-molecule drug discovery in SMA, identifies a novel mechanism of action for an E7 splicing modifier, and sets a precedent for other splicing-mediated diseases where RNA structure could be similarly targeted.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-SMN antibody, Mouse monoclonal, clone 2B1, purified from hybridoma cell culture
Sigma-Aldrich
PK4C9, ≥98% (HPLC)