
Xanthanoic acid

Synonym(s):
Xanthene-9-carboxylic acid
Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2