
Wortmannin

Synonym(s):
KY 12420, Wortmannin, Antibiotic SL-2052
Empirical Formula (Hill Notation):
C23H24O8
CAS Number:
Molecular Weight:
428.43
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment