
Tubocurarine chloride

Synonym(s):
Tubocurarine hydrochloride pentahydrate, (+)-Tubocurarine hydrochloride
Empirical Formula (Hill Notation):
C37H41ClN2O6 · HCl · 5H2O
CAS Number:
Molecular Weight:
771.72
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing