
Triton™ X-114

Synonym(s):
(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, Polyethylene glycol tert-octylphenyl ether
Linear Formula:
(C2H4O)n C14H22O, n = 7 or 8
CAS Number:
Products
Product Category
Brand
Feature
Physical Form
Chemical Composition
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing