
Triton™ X-100 reduced

Synonym(s):
Polyoxyethylene (10) isooctylcyclohexyl ether
Linear Formula:
4-(C8H17)C6H10(OCH2CH2)nOH, n~10
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing