
Testosterone Related Compound C

Synonym(s):
Epitestosterone, 17α-Hydroxy-4-androsten-3-one, 4-Androsten-17α-ol-3-one, cis-Testosterone, Isotestosterone
Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing