
Testosterone for ID and assay

Synonym(s):
Testosterone, 17β-Hydroxy-3-oxo-4-androstene, 17β-Hydroxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one, trans-Testosterone
Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
Products
Formula Weight
Gene Alias
Manufacturer
Quality Segment
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing