(S,S)-(+)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine
Synonym(s):
(S,S)-Jacobsen’s ligand
Linear Formula:
[[(CH3)3C]2C6H2-2-(OH)CH=N]2C6H10
CAS Number:
Molecular Weight:
546.83
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing