
Carbon disulfide solution

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Products
Brand
Formula Weight
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3