
Salbutamol impurity I

Synonym(s):
(1RS)-2-[(1,1-Dimethylethyl)amino]-1-[3-(hydroxymethyl)-4-benzyloxyphenyl]ethanol, 2-tert-Butylamino-1-[3-(hydroxymethyl)-4-benzyloxyphenyl]ethanol
Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2