Skip to Content
Merck
R-Methotrexate certified reference material, pharmaceutical secondary standard

R-Methotrexate

R-Methotrexate certified reference material, pharmaceutical secondary standard
Synonym(s):
(−)-Amethopterin
Empirical Formula (Hill Notation):
C20H22N8O5
CAS Number:
Molecular Weight:
454.44

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade