
Retinyl Acetate-Water Soluble

Synonym(s):
Retinyl acetate, Retinol acetate, Vitamin A acetate
Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing