Quinidine sulfate
Synonym(s):
Quinidine sulfate salt dihydrate
Empirical Formula (Hill Notation):
C40H48N4O4 · H2O4S · 2H2O
CAS Number:
Molecular Weight:
782.94
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade