
Psoralidin

Synonym(s):
3,9-Dihydroxy-2-(3-methyl-2-butenyl)-6H-benzofuro[3,2-c][1]benzopyran-6-one, 3,9-Dihydroxy-2-prenylcoumestan
Empirical Formula (Hill Notation):
C20H16O5
CAS Number:
Molecular Weight:
336.34
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing