
Polyinosinic–polycytidylic acid sodium salt

Synonym(s):
Poly (I:C), Poly(I) • Poly(C)
CAS Number:
Products
Brand
Biological Source
Physical Form
Purity
Quality Segment
Sterilization Method
Showing 1-2 of 2
Compare
Product Number
Description
Pricing