
Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid

Synonym(s):
mercaptoPEG acid, mercaptopoly(ethylene glycol) carboxylic acid, mercaptopolyoxyethylene acetic acid, thio-PEG, thiol-PEG-acid, thiol-PEG-carboxylate
Linear Formula:
HSC2H4O(C2H4O)nCH2CO2H
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-4 of 4
Compare
Product Number
Description
Pricing