Skip to Content
Merck
Phosphorus standard solution 1000 ppm P

Phosphorus standard solution

Phosphorus standard solution 1000 ppm P
Synonym(s):
Potassium Phosphate Monobasic solution, Monopotassium phosphate, Potassium dihydrogen phosphate solution
CAS Number:

Products

Product Category
Brand
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing