
Phorbol 12-myristate 13-acetate

Synonym(s):
12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA, Phorbol-12-myristate-13-acetate, InSolution Phorbol-12-myristate-13-acetate
Empirical Formula (Hill Notation):
C36H56O8
CAS Number:
Molecular Weight:
616.83
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-4 of 4
Compare
Product Number
Description
Pricing