
N-Methyl-N-perfluorooctylsulfonylaminoethanol

Synonym(s):
1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-(2-hydroxyethyl)-N-methyl-1-octanesulfonamide, N-Methylperfluorooctanesulfonamidoethanol
Empirical Formula (Hill Notation):
C11H8F17NO3S
CAS Number:
Molecular Weight:
557.22
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing