
N-Boc-1,6-hexanediamine hydrochloride

Synonym(s):
N-Boc-1,6-diamino-hexane hydrochloride, tert-Butyl N-(6-aminohexyl)carbamate hydrochloride
Linear Formula:
(CH3)3COCONH(CH2)6NH2 · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing