
Naloxone hydrochloride dihydrate

Synonym(s):
Naloxone hydrochloride dihydrate
Empirical Formula (Hill Notation):
C19H21NO4 · HCl · 2H2O
CAS Number:
Molecular Weight:
399.87
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing