![Methyl γ-linolenate ≥99% (GC)](/deepweb/assets/sigmaaldrich/product/structures/116/606/a291811d-0f6f-4a49-bd7b-44d6a213acd4/640/a291811d-0f6f-4a49-bd7b-44d6a213acd4.png)
Methyl γ-linolenate
![Methyl γ-linolenate ≥99% (GC)](/deepweb/assets/sigmaaldrich/product/structures/116/606/a291811d-0f6f-4a49-bd7b-44d6a213acd4/640/a291811d-0f6f-4a49-bd7b-44d6a213acd4.png)
Synonym(s):
γ-Linolenic acid methyl ester, ω-6 Linolenic acid methyl ester, 6,9,12-Octadecatrienoic acid methyl ester, Fame 18:3n-6, Methyl (Z,Z,Z)-6,9,12-octadecatrienoate
Linear Formula:
CH3(CH2)3(CH2CH=CH)3(CH2)4CO2CH3
CAS Number:
Molecular Weight:
292.46
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing