
Methacrylic acid copolymer type B

Synonym(s):
Poly(methyl methacrylate-co-methacrylic acid), Poly(methacrylic acid-co-methyl methacrylate)
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing