
Mebeverine hydrochloride

Synonym(s):
Mebeverine hydrochloride
Empirical Formula (Hill Notation):
C25H35NO5 · HCl
CAS Number:
Molecular Weight:
466.01
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-2 of 2