
Manganese(II) phthalocyanine

Synonym(s):
Phthalocyanine manganese(II) salt
Empirical Formula (Hill Notation):
C32H16MnN8
CAS Number:
Molecular Weight:
567.46
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing