Skip to Content
Merck
Lidocaine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Lidocaine Related Compound A

Lidocaine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2,6-Dimethylaniline, 2,6-Xylidine, 2-Amino-1,3-dimethylbenzene, 2-Amino-m-xylene
Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1