L-Glutamic acid
Synonym(s):
(S)-2-Aminopentanedioic acid, Glu, L-Glutamic acid
Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
Reference Material Type
Special Grade
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing