Skip to Content
Merck
Isooctane for analysis EMPARTA® ACS

Isooctane

Isooctane for analysis EMPARTA® ACS
Synonym(s):
2,2,4-Trimethylpentane, Isooctane
Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Quality Segment
Technique