
Isoflurane Related Compound B

Synonym(s):
Difluoromethyl 2,2,2-trifluoroethyl ether, 2,2,2-Trifluoroethyl difluoromethyl ether
Empirical Formula (Hill Notation):
C3H3F5O
CAS Number:
Molecular Weight:
150.05
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2