![Hymecromone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/313/985/b4aed9cd-da02-4393-a37b-64379aac11f0/640/b4aed9cd-da02-4393-a37b-64379aac11f0.png)
Hymecromone
![Hymecromone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/313/985/b4aed9cd-da02-4393-a37b-64379aac11f0/640/b4aed9cd-da02-4393-a37b-64379aac11f0.png)
Synonym(s):
4-Methylumbelliferone, β-Methylumbelliferone, 4-MU, 7-Hydroxy-4-methylcoumarin
Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing