
Fmoc-D-Phe(4-CN)-OH

Synonym(s):
Fmoc-4-cyano-D-phenylalanine
Empirical Formula (Hill Notation):
C25H20N2O4
CAS Number:
Molecular Weight:
412.44
Products
Product Category
Brand
Formula Weight
Physical Form
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing