
Ethylene glycol dimethyl ether

Synonym(s):
1,2-Dimethoxyethane, mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme
Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing