
Ethisterone

Synonym(s):
17α-Ethynyltestosterone, 17β-Hydroxy-4,17α-pregnen-20-yn-3-one, 4,17α-Pregnen-17β-ol-20-yn-3-one
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Sterility
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing