
Etacrynic acid for system suitability

Synonym(s):
Ethacrynic acid, (2,3-Dichloro-4-[2-methylenebutyryl]phenoxy)acetic acid
Empirical Formula (Hill Notation):
C13H12Cl2O4
CAS Number:
Molecular Weight:
303.14
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing