
(±)-Sulpiride

Synonym(s):
(±)-5-(Aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide, (±)-N-1-(Ethylpyrrolidin-2-ylmethyl)-2-methoxy-5-sulfamoylbenzamide
Empirical Formula (Hill Notation):
C15H23N3O4S
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing